Assay Details
| Gene Symbol: Hdgfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133549.2 | NP_598233.2 | 1 | 1764 | 175 | |
| GenBank mRNA | AY061636.1 | - | 2 | 1355 | 174 | |
| BC085707.1 | - | 1 | 1764 | 7132556 | 175 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hdgfl1
Genomic Map