Assay Details
| Gene Symbol: Acot13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106111.1 | NP_001099581.1 | 1 - 2 | 314 | 173 | |
| GenBank mRNA | FQ217526.1 | - | 1 - 2 | 264 | 173 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106111.1 transcripts.
Set Membership: |
|

Acot13
Genomic Map