Assay Details
| Gene Symbol: Fopnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108261.1 | NP_001101731.1 | 1 - 2 | 70 | 105 | |
| XM_006245848.2 | XP_006245910.1 | 1 - 2 | 99 | 105 | ||
| GenBank mRNA | BC083733.1 | - | 1 - 2 | 44 | 105 | |
| FQ228774.1 | - | 1 - 2 | 44 | 105 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001108261.1,XM_006245848.2 transcripts.
This assay is in the 5'UTR region of the NM_001108261.1,XM_006245848.2 transcripts.
Set Membership: |
|

Fopnl
Genomic Map