Assay Details
| Gene Symbol: Fopnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108261.1 | NP_001101731.1 | 2 - 3 | 280 | 140 | |
| XM_006245848.2 | XP_006245910.1 | 2 - 3 | 309 | 140 | ||
| GenBank mRNA | BC083733.1 | - | 2 - 3 | 254 | 140 | |
| BC158662.1 | - | 2 - 3 | 246 | 8369763 | 140 | |
| FQ215640.1 | - | 2 - 3 | 246 | 140 | ||
| FQ228774.1 | - | 2 - 3 | 254 | 140 | ||
| FQ228837.1 | - | 2 - 3 | 236 | 140 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fopnl
Genomic Map