Assay Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766411.1 | XP_008764633.1 | 3 - 4 | 1173 | 125 | |
| XM_343420.8 | XP_343421.3 | 3 - 4 | 1165 | 125 | ||
More Information
Set Membership: |
|

Fam63b
Genomic Map