Assay Details
| Gene Symbol: Zfp280d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108165.2 | NP_001101635.2 | 16 - 17 | 2132 | 116 | |
| XM_006243356.2 | XP_006243418.1 | 15 - 16 | 2099 | 116 | ||
| XM_006243357.2 | XP_006243419.1 | 16 - 17 | 2095 | 116 | ||
| XM_008766288.1 | XP_008764510.1 | 15 - 16 | 2082 | 116 | ||
| XM_008766289.1 | XP_008764511.1 | 17 - 18 | 2275 | 116 | ||
| XM_008766290.1 | XP_008764512.1 | 15 - 16 | 2082 | 116 | ||
| GenBank mRNA | FQ222707.1 | - | 17 - 18 | 2280 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp280d
Genomic Map