Assay Details
| Gene Symbol: Ice2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191108.1 | NP_001178037.1 | 14 - 15 | 2866 | 52 | |
| XM_006243402.2 | XP_006243464.1 | 15 - 16 | 3334 | 52 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ice2
Genomic Map