Assay Details
| Gene Symbol: Lipc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012597.2 | NP_036729.2 | 1 - 2 | 181 | 105 | |
| XM_006243359.2 | XP_006243421.1 | 2 - 3 | 236 | 105 | ||
| XM_006243360.2 | XP_006243422.1 | 2 - 3 | 236 | 105 | ||
| XM_006243361.2 | XP_006243423.1 | 2 - 3 | 236 | 105 | ||
| GenBank mRNA | BC088160.1 | - | 1 - 2 | 181 | 7323251 | 105 |
| M16235.1 | - | 1 - 2 | 103 | 105 | ||
| X17366.1 | - | 1 - 2 | 105 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Lipc
Genomic Map