Assay Details
| Gene Symbol: LOC103689982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758837.1 | XP_008757059.1 | 3 - 4 | 428 | 69 | |
| Gene Symbol: Mboat7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134978.1 | NP_001128450.1 | 428 | 69 | ||
| GenBank mRNA | BC169089.1 | - | 428 | 5598892 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103689982
Genomic Map