Assay Details
| Gene Symbol: Idua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172084.1 | NP_001165555.1 | 7 - 8 | 1053 | 62 | |
| XM_008769938.1 | XP_008768160.1 | 5 - 6 | 691 | 62 | ||
| GenBank mRNA | BC083854.1 | - | 7 - 8 | 889 | 62 | |
| BC103733.1 | - | 6 - 7 | 1066 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Idua
Genomic Map