Assay Details
| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002831.1 | NP_001002831.1 | 3 - 4 | 245 | 83 | |
| GenBank mRNA | FQ209948.1 | - | 2 - 3 | 277 | 83 | |
| FQ211619.1 | - | 3 - 4 | 266 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpp21
Genomic Map