Assay Details
| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002831.1 | NP_001002831.1 | 1 - 2 | 59 | 85 | |
| GenBank mRNA | FQ211619.1 | - | 1 - 2 | 80 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpp21
Genomic Map