Assay Details
| Gene Symbol: RT1-M3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022921.2 | NP_075210.2 | 4 - 5 | 925 | 142 | |
| XM_008772688.1 | XP_008770910.1 | 4 - 5 | 936 | 142 | ||
| XM_008772689.1 | XP_008770911.1 | 4 - 5 | 936 | 142 | ||
| GenBank mRNA | AJ249342.1 | - | 4 - 5 | 924 | 142 | |
| U16025.1 | - | 4 - 5 | 898 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-M3-1
Genomic Map