Assay Details
| Gene Symbol: Znrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166300.1 | NP_001159772.1 | 2 - 3 | 246 | 92 | |
| NM_213567.2 | NP_998732.1 | 1 - 2 | 164 | 92 | ||
| GenBank mRNA | AI232728.1 | - | 3 - 4 | 92 | ||
| FM113828.1 | - | 2 - 3 | 245 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Znrd1
Genomic Map