Assay Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133387.2 | NP_596878.2 | 7 - 8 | 1109 | 163 | |
| XM_006255855.1 | XP_006255917.1 | 6 - 7 | 1118 | 163 | ||
| GenBank mRNA | AF374406.1 | - | 5 - 6 | 950 | 163 | |
| BC085923.1 | - | 7 - 8 | 1109 | 7109209 | 163 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmlhe
Genomic Map