Assay Details
| Gene Symbol: Acrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021747.1 | NP_068515.1 | 2 - 3 | 577 | 148 | |
| GenBank mRNA | AJ243484.1 | - | 2 - 3 | 577 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acrv1
Genomic Map