Assay Details
| Gene Symbol: Wbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257281.2 | XP_006257343.1 | 2 - 3 | 216 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257281.2 transcripts.
Set Membership: |
|

Wbp5
Genomic Map