Assay Details
| Gene Symbol: Bhlhb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257276.2 | XP_006257338.1 | 3 - 4 | 1464 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257276.2 transcripts.
Set Membership: |
|

Bhlhb9
Genomic Map