Assay Details
| Gene Symbol: Bhlhb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257273.2 | XP_006257335.1 | 3 - 4 | 2603 | 90 | |
| XM_006257274.2 | XP_006257336.1 | 4 - 5 | 2454 | 90 | ||
| XM_006257275.2 | XP_006257337.1 | 3 - 4 | 1417 | 90 | ||
| XM_006257276.2 | XP_006257338.1 | 4 - 5 | 1565 | 90 | ||
| XM_008773421.1 | XP_008771643.1 | 3 - 4 | 1399 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257273.2, XM_006257274.2, XM_006257275.2, XM_006257276.2, XM_008773421.1 transcripts.
Set Membership: |
|

Bhlhb9
Genomic Map