Assay Details
| Gene Symbol: Mt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126084.1 | NP_001119556.1 | 1 - 2 | 62 | 101 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001126084.1 transcripts.
Set Membership: |
|

Mt4
Genomic Map