Assay Details
| Gene Symbol: Ces2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144743.1 | NP_653344.1 | 829 | 99 | ||
| GenBank mRNA | AY034877.1 | - | 829 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces2a
Genomic Map