Assay Details
| Gene Symbol: Ces2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144743.1 | NP_653344.1 | 924 | 93 | ||
| GenBank mRNA | AY034877.1 | - | 924 | 93 | ||
| Gene Symbol: Ces2j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190380.1 | NP_001177309.1 | 905 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ces2a
Genomic Map