Assay Details
| Gene Symbol: Thoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047850.1 | NP_001041315.1 | 7 - 8 | 652 | 79 | |
| XM_006254399.2 | XP_006254461.1 | 11 - 12 | 1148 | 79 | ||
| GenBank mRNA | AY387056.1 | - | 7 - 8 | 652 | 79 | |
| FQ229414.1 | - | 1 - 2 | 94 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thoc1
Genomic Map