Assay Details
| Gene Symbol: RT1-S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008886.2 | NP_001008886.2 | 1 - 2 | 105 | 109 | |
| XM_006255917.2 | XP_006255979.1 | 1 - 2 | 215 | 109 | ||
| GenBank mRNA | AF029241.1 | - | 1 - 2 | 30 | 109 | |
| CK599963.1 | - | 1 - 2 | 105 | 109 | ||
| FQ209394.1 | - | 1 - 2 | 94 | 109 | ||
| FQ232669.1 | - | 1 - 2 | 93 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-S3
Genomic Map