Assay Details
| Gene Symbol: Abcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109883.2 | NP_001103353.1 | 8 - 9 | 706 | 69 | |
| GenBank mRNA | AF293383.1 | - | 7 - 8 | 602 | 69 | |
| CK356298.1 | - | 2 - 3 | 188 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcf1
Genomic Map