Assay Details
| Gene Symbol: Cb707485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029398.1 | 1 - 2 | 102 | 64 | ||
| GenBank mRNA | CB707485.1 | - | 1 - 2 | 115 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cb707485
Genomic Map