Assay Details
| Gene Symbol: Pou5f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009178.2 | NP_001009178.1 | 4 - 5 | 881 | 68 | |
| GenBank mRNA | BC158566.1 | - | 4 - 5 | 881 | 9092936 | 68 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pou5f1
Genomic Map