Assay Details
| Gene Symbol: RGD1302996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213610.3 | NP_998775.2 | 3 - 4 | 623 | 106 | |
| XM_006255918.2 | XP_006255980.1 | 3 - 4 | 641 | 106 | ||
| XM_008772712.1 | XP_008770934.1 | 3 - 4 | 512 | 106 | ||
| XM_008772713.1 | XP_008770935.1 | 3 - 4 | 500 | 106 | ||
| GenBank mRNA | BC088194.1 | - | 3 - 4 | 623 | 7373688 | 106 |
| FQ215898.1 | - | 3 - 4 | 628 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1302996
Genomic Map