Assay Details
| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106793.2 | NP_001100263.2 | 13 - 14 | 1854 | 76 | |
| XM_006257343.2 | XP_006257405.1 | 13 - 14 | 1943 | 76 | ||
| GenBank mRNA | BC169082.1 | - | 12 - 13 | 1733 | 76 | |
| CO570626.1 | - | 3 - 4 | 488 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx23
Genomic Map