Assay Details
| Gene Symbol: Dbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001053826.4 | XP_001053826.1 | 3 - 4 | 864 | 77 | |
| XM_006242286.2 | XP_006242348.1 | 2 - 3 | 768 | 77 | ||
| GenBank mRNA | AB121147.1 | - | 1 - 2 | 170 | 77 | |
More Information
Set Membership: |
|

Dbx2
Genomic Map