Assay Details
| Gene Symbol: Tuba1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022298.1 | NP_071634.1 | 1 - 2 | 71 | 73 | |
| GenBank mRNA | BC062238.1 | - | 1 - 2 | 105 | 6922126 | 73 |
| BC078830.1 | - | 1 - 2 | 84 | 7115494 | 73 | |
| FQ210110.1 | - | 1 - 2 | 103 | 73 | ||
| FQ213186.1 | - | 1 - 2 | 103 | 73 | ||
| V01227.1 | - | 1 - 2 | 71 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062238.1,BC078830.1,NM_022298.1,V01227.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tuba1a
Genomic Map