Assay Details
| Gene Symbol: Krt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008899.1 | NP_001008899.1 | 6 - 7 | 1305 | 93 | |
| XM_006242436.2 | XP_006242498.1 | 6 - 7 | 1806 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt2
Genomic Map