Assay Details
| Gene Symbol: Krt84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008812.1 | NP_001008812.1 | 5 - 6 | 1120 | 147 | |
| XM_006242346.2 | XP_006242408.1 | 5 - 6 | 1189 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt84
Genomic Map