Assay Details
| Gene Symbol: Rarg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135249.1 | NP_001128721.1 | 8 - 9 | 1501 | 67 | |
| NM_001135250.1 | NP_001128722.1 | 6 - 7 | 1273 | 67 | ||
| XM_008765784.1 | XP_008764006.1 | 7 - 8 | 1474 | 67 | ||
| XM_008765785.1 | XP_008764007.1 | 8 - 9 | 1465 | 67 | ||
| XM_008765786.1 | XP_008764008.1 | 7 - 8 | 1993 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rarg
Genomic Map