Assay Details
| Gene Symbol: Krt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053976.1 | NP_446428.1 | 5 - 6 | 961 | 108 | |
| GenBank mRNA | BC091275.1 | - | 5 - 6 | 961 | 7383311 | 108 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krt18
Genomic Map