Assay Details
| Gene Symbol: Riiad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144957.1 | NP_001138429.1 | 3 - 4 | 204 | 100 | |
| GenBank mRNA | CO401253.1 | - | 3 - 4 | 168 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riiad1
Genomic Map