Assay Details
| Gene Symbol: Rhoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106461.1 | NP_001099931.1 | 2 - 3 | 422 | 93 | |
| XM_006233074.1 | XP_006233136.1 | 2 - 3 | 396 | 93 | ||
| GenBank mRNA | BC166849.1 | - | 2 - 3 | 438 | 9088252 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|

Rhoc
Genomic Map