Assay Details
| Gene Symbol: Hipk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100986.1 | NP_001094456.1 | 10 - 11 | 2423 | 75 | |
| XM_006233090.2 | XP_006233152.1 | 10 - 11 | 2374 | 75 | ||
| XM_006233097.2 | XP_006233159.1 | 10 - 11 | 2271 | 75 | ||
| XM_008761412.1 | XP_008759634.1 | 10 - 11 | 2735 | 75 | ||
| XM_008761413.1 | XP_008759635.1 | 10 - 11 | 3434 | 75 | ||
| XM_008761414.1 | XP_008759636.1 | 11 - 12 | 3589 | 75 | ||
| XM_008761415.1 | XP_008759637.1 | 11 - 12 | 3920 | 75 | ||
| XM_008761416.1 | XP_008759638.1 | 11 - 12 | 3259 | 75 | ||
| XM_008761417.1 | XP_008759639.1 | 10 - 11 | 2937 | 75 | ||
| XM_008761418.1 | XP_008759640.1 | 10 - 11 | 3039 | 75 | ||
| XM_008761419.1 | XP_008759641.1 | 10 - 11 | 2736 | 75 | ||
| XM_008761420.1 | XP_008759642.1 | 10 - 11 | 2976 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hipk1
Genomic Map