Assay Details
| Gene Symbol: Scnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107696.1 | NP_001101166.1 | 1 - 2 | 165 | 86 | |
| GenBank mRNA | FQ232097.1 | - | 123 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scnm1
Genomic Map