Assay Details
| Gene Symbol: Scnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107696.1 | NP_001101166.1 | 5 - 6 | 505 | 85 | |
| XM_006232882.2 | XP_006232944.1 | 4 - 5 | 344 | 85 | ||
| XM_006232883.2 | XP_006232945.1 | 5 - 6 | 423 | 85 | ||
| GenBank mRNA | FQ232097.1 | - | 463 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scnm1
Genomic Map