Assay Details
| Gene Symbol: Dennd2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191569.1 | NP_001178498.1 | 14 - 15 | 2558 | 65 | |
| XM_006233069.1 | XP_006233131.1 | 14 - 15 | 2549 | 65 | ||
| XM_006233070.2 | XP_006233132.1 | 14 - 15 | 2876 | 65 | ||
| XM_006233071.2 | XP_006233133.1 | 14 - 15 | 2661 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2c
Genomic Map