Assay Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134593.1 | NP_001128065.1 | 5 - 6 | 763 | 67 | |
| XM_008761317.1 | XP_008759539.1 | 10 - 11 | 3234 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ciart
Genomic Map