Assay Details
| Gene Symbol: Sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011951.1 | NP_001011951.1 | 3 - 4 | 946 | 56 | |
| GenBank mRNA | BC078997.1 | - | 3 - 4 | 946 | 7112738 | 56 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sf3b4
Genomic Map