Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109625.2 | NP_001103095.2 | 2 - 3 | 388 | 129 | |
| GenBank mRNA | BC166464.1 | - | 2 - 3 | 103 | 9030793 | 129 |
| EV765154.1 | - | 2 - 3 | 388 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capza1
Genomic Map