Assay Details
| Gene Symbol: Aph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014255.1 | NP_001014277.1 | 2 - 3 | 473 | 105 | |
| XM_006232977.2 | XP_006233039.1 | 2 - 3 | 526 | 105 | ||
| GenBank mRNA | BC087081.1 | - | 2 - 3 | 473 | 7189126 | 105 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aph1a
Genomic Map