Assay Details
| Gene Symbol: Prune | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007697.1 | NP_001007698.1 | 1 - 2 | 294 | 93 | |
| XM_006232885.2 | XP_006232947.1 | 1 - 2 | 306 | 93 | ||
| GenBank mRNA | BC079054.1 | - | 1 - 2 | 294 | 7115417 | 93 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC079054.1,NM_001007697.1,XM_006232885.2 transcripts.
Set Membership: |
|

Prune
Genomic Map