Assay Details
| Gene Symbol: Prune | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007697.1 | NP_001007698.1 | 2 - 3 | 389 | 144 | |
| XM_006232885.2 | XP_006232947.1 | 2 - 3 | 401 | 144 | ||
| GenBank mRNA | BC079054.1 | - | 2 - 3 | 389 | 7115417 | 144 |
More Information
Updated Mapping Information
Set Membership: |
|

Prune
Genomic Map