Assay Details
| Gene Symbol: LOC100363520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232939.2 | XP_006233001.1 | 4 - 5 | 804 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100363520| Gene Symbol: LOC100363520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232939.2 | XP_006233001.1 | 4 - 5 | 804 | 62 | |
Set Membership: |
|