Assay Details
| Gene Symbol: Hmbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252219.2 | XP_006252281.1 | 7 - 8 | 1268 | 63 | |
| XM_006252220.2 | XP_006252282.1 | 7 - 8 | 1268 | 63 | ||
| XM_006252221.2 | XP_006252283.1 | 7 - 8 | 1267 | 63 | ||
| XM_008770802.1 | XP_008769024.1 | 7 - 8 | 1195 | 63 | ||
| XM_008770803.1 | XP_008769025.1 | 7 - 8 | 1268 | 63 | ||
| XM_008770804.1 | XP_008769026.1 | 7 - 8 | 1268 | 63 | ||
| XM_008770805.1 | XP_008769027.1 | 7 - 8 | 1268 | 63 | ||
More Information
Set Membership: |
|

Hmbox1
Genomic Map