Assay Details
| Gene Symbol: Dpysl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105717.2 | NP_001099187.1 | 11 - 12 | 1503 | 73 | |
| XM_006252097.2 | XP_006252159.1 | 11 - 12 | 1734 | 73 | ||
| GenBank mRNA | CA510768.1 | - | 5 - 6 | 620 | 73 | |
| CB577285.1 | - | 2 - 3 | 251 | 73 | ||
| Z46882.1 | - | 11 - 12 | 1456 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to amphetamine
pyrimidine nucleobase catabolic process
endocytosis
cytoskeleton organization
brain development
regulation of neuron projection development
positive regulation of glutamate secretion
spinal cord development
olfactory bulb development
regulation of axon extension
response to cocaine
response to drug
regulation of neuron differentiation
synaptic vesicle transport
protein binding
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amides
protein kinase binding
response to amphetamine
pyrimidine nucleobase catabolic process
endocytosis
cytoskeleton organization
brain development
regulation of neuron projection development
positive regulation of glutamate secretion
spinal cord development
olfactory bulb development
regulation of axon extension
response to cocaine
response to drug
regulation of neuron differentiation
synaptic vesicle transport
protein binding
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amides
protein kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P311647 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77750 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15655 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P311647 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P311647 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15655 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15655 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P311647 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P271741 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P311647 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702155811 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21644 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9681 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702155811 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10784693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784688 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784685 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784689 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784690 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784680 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784687 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10784679 | - | - |
| Rat Genome 230 2.0 Array | 1371694_at | - | - |
| Rat Genome 230 2.0 Array | 1371694_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380728_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380728_at | - | - |
| Affymetrix HT Rat Focus Array | 1371694_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371694_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500012545.rn.1 | TC1500001866.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10784679 | - | - |
| Rat Genome U34 Set | Z46882cds_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21849033 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355161 | - | - |

Dpysl2
Genomic Map